Order Entry
Sweden
ContactUsLinkComponent
Muffle Furnace L3/12/C550 Tmax1200 with flap door and protective gas connection, over-temperature limiter and thermocouple port in t he rear wall 1 * 1 items
Katalog # 455-2586
Supplier:  NABERTHERM
New Product
Muffle Furnace L3/12/C550 Tmax1200 with flap door and protective gas connection, over-temperature limiter and thermocouple port in t he rear wall 1 * 1 items
Katalog # 455-2586
Supplier:  NABERTHERM