Heliport Plus Heated Rack Thawing Station - The Heliport Plus heated rack thawing station thaws frozen samples and reagents in a fra ction of the time. A heating element can be turned on when needed, for samples stored in DMSO and turned off for aque 1 * 1 Items
Catalog # MSPPJDM1-300-71000
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together