HeliPad Large tube Rack The new HeliPad racks can be placed anywhere on the Heliport deck and can be mixed with other rack consumabl es. The small HeliPad rack has 40 holes for 2 ml size tubes (11mm). The large HeliPad rack has 14 medium size holes ( 1 * 1 Items
Catalog # MSPPJDM1-300-82000
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together