Order Entry
Sweden
ContactUsLinkComponent
Human IL-2 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant human IL-2 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 576 Tests
  ANTIA270489-696
New Product
Human IL-2 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant human IL-2 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 576 Tests
  ANTIA270489-696
New Product