Order Entry
Sweden
ContactUsLinkComponent
Monkey IgA Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled polyclonal detection antibody , Streptavidin-AP, native human IgA ELISA standard, and standard reconstitution buffer. This matched antibody pair ki 1 * 576 Tests
Catalog # ANTIA270425-696
Supplier:  ANTIBODIES.COM
New Product
Monkey IgA Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled polyclonal detection antibody , Streptavidin-AP, native human IgA ELISA standard, and standard reconstitution buffer. This matched antibody pair ki 1 * 576 Tests
Catalog # ANTIA270425-696
Supplier:  ANTIBODIES.COM
New Product