Order Entry
Sweden
ContactUsLinkComponent
Bovine IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an HRP-labelled monoclonal detection antibod y, recombinant bovine IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can be u 1 * 576 Tests
Catalog # ANTIA270458-696
Supplier:  ANTIBODIES.COM
New Product
Bovine IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an HRP-labelled monoclonal detection antibod y, recombinant bovine IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can be u 1 * 576 Tests
Catalog # ANTIA270458-696
Supplier:  ANTIBODIES.COM
New Product