Order Entry
Sweden
ContactUsLinkComponent
Mouse IL-6 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-HRP, recombinant mouse IL-6 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 1.920 Tests
Catalog # ANTIA270528-2096
Supplier:  Antibodies.com
New Product
Mouse IL-6 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-HRP, recombinant mouse IL-6 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 1.920 Tests
Catalog # ANTIA270528-2096
Supplier:  Antibodies.com