Order Entry
Sweden
ContactUsLinkComponent
Human Thioredoxin/TRX Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detecti on antibody, Streptavidin-AP, recombinant human thioredoxin-1 ELISA standard, and standard reconstitution buffer. Thi 1 * 576 Tests
Catalog # ANTIA270420-696
Supplier:  ANTIBODIES.COM
New Product
Human Thioredoxin/TRX Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detecti on antibody, Streptavidin-AP, recombinant human thioredoxin-1 ELISA standard, and standard reconstitution buffer. Thi 1 * 576 Tests
Catalog # ANTIA270420-696
Supplier:  ANTIBODIES.COM
New Product