Order Entry
Sweden
ContactUsLinkComponent
Porcine IL-2 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-AP, recombinant porcine IL-2 ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Catalog # ANTIA270450-2096
Supplier:  ANTIBODIES.COM
New Product
Porcine IL-2 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-AP, recombinant porcine IL-2 ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Catalog # ANTIA270450-2096
Supplier:  ANTIBODIES.COM
New Product