Order Entry
Sweden
ContactUsLinkComponent
Porcine TNF alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection a ntibody, Streptavidin-HRP, recombinant porcine TNF alpha ELISA standard, and standard reconstitution buffer. This mat 1 * 1.920 Tests
Catalog # ANTIA270534-2096
Supplier:  ANTIBODIES.COM
New Product
Porcine TNF alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection a ntibody, Streptavidin-HRP, recombinant porcine TNF alpha ELISA standard, and standard reconstitution buffer. This mat 1 * 1.920 Tests
Catalog # ANTIA270534-2096
Supplier:  ANTIBODIES.COM
New Product