Order Entry
Sweden
ContactUsLinkComponent
Human Perforin Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection anti body, Streptavidin-AP, native human perforin ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 576 Tests
Catalog # ANTIA270418-696
Supplier:  ANTIBODIES.COM
New Product
Human Perforin Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection anti body, Streptavidin-AP, native human perforin ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 576 Tests
Catalog # ANTIA270418-696
Supplier:  ANTIBODIES.COM
New Product