Order Entry
Sweden
ContactUsLinkComponent
Monkey IgA Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled polyclonal detection antibody , Streptavidin-HRP, native human IgA ELISA standard, and standard reconstitution buffer. This matched antibody pair k 1 * 576 Tests
  ANTIA270509-696
New Product
Monkey IgA Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled polyclonal detection antibody , Streptavidin-HRP, native human IgA ELISA standard, and standard reconstitution buffer. This matched antibody pair k 1 * 576 Tests
  ANTIA270509-696
New Product