Order Entry
Sweden
ContactUsLinkComponent
Ferret IL-2 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant porcine IL-2 ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Catalog # ANTIA270469-2096
Supplier:  ANTIBODIES.COM
New Product
Ferret IL-2 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant porcine IL-2 ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Catalog # ANTIA270469-2096
Supplier:  ANTIBODIES.COM
New Product