Order Entry
Sweden
ContactUsLinkComponent
Human IL-8 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-HRP, recombinant human IL-8 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 1.920 Tests
  ANTIA270500-2096
New Product
Human IL-8 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-HRP, recombinant human IL-8 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 1.920 Tests
  ANTIA270500-2096
New Product