Order Entry
Sweden
ContactUsLinkComponent
Human Granzyme A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant human granzyme A ELISA standard, and standard reconstitution buffer. This matche 1 * 1.920 Tests
Catalog # ANTIA270390-2096
Supplier:  ANTIBODIES.COM
New Product
Human Granzyme A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant human granzyme A ELISA standard, and standard reconstitution buffer. This matche 1 * 1.920 Tests
Catalog # ANTIA270390-2096
Supplier:  ANTIBODIES.COM
New Product