Order Entry
Sweden
ContactUsLinkComponent
Human IL-5 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant human IL-5 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 1.920 Tests
Catalog # ANTIA270414-2096
Supplier:  Antibodies.com
New Product
Human IL-5 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant human IL-5 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 1.920 Tests
Catalog # ANTIA270414-2096
Supplier:  Antibodies.com