Order Entry
Sweden
ContactUsLinkComponent
Human IL-29 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant human IL-29 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
  ANTIA270410-696
New Product
Human IL-29 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant human IL-29 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
  ANTIA270410-696
New Product