Order Entry
Sweden
ContactUsLinkComponent
Human Perforin Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection anti body, Streptavidin-AP, native human perforin ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 1.920 Tests
  ANTIA270418-2096
New Product
Human Perforin Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection anti body, Streptavidin-AP, native human perforin ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 1.920 Tests
  ANTIA270418-2096
New Product