Order Entry
Sweden
ContactUsLinkComponent
Human IL-4 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant human IL-4 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 576 Tests
Catalog # ANTIA270413-696
Supplier:  ANTIBODIES.COM
New Product
Human IL-4 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant human IL-4 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 576 Tests
Catalog # ANTIA270413-696
Supplier:  ANTIBODIES.COM
New Product