Order Entry
Sweden
ContactUsLinkComponent
Rat Complement C3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of rat Complement C3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 48 Tests
Catalog # ANTIA2004-48
Leverantör:  ANTIBODIES.COM
New Product
Rat Complement C3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of rat Complement C3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 48 Tests
Catalog # ANTIA2004-48
Leverantör:  ANTIBODIES.COM
New Product