Order Entry
Sweden
ContactUsLinkComponent
Monkey TNF alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant human TNF alpha ELISA standard, and standard reconstitution buffer. This matched 1 * 576 Tests
Catalog # ANTIA270430-696
Supplier:  ANTIBODIES.COM
New Product
Monkey TNF alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant human TNF alpha ELISA standard, and standard reconstitution buffer. This matched 1 * 576 Tests
Catalog # ANTIA270430-696
Supplier:  ANTIBODIES.COM
New Product