Order Entry
Sweden
ContactUsLinkComponent
Human Granzyme B Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant human granzyme B ELISA standard, and standard reconstitution buffer. This matche 1 * 576 Tests
Catalog # ANTIA270391-696
Supplier:  ANTIBODIES.COM
New Product
Human Granzyme B Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant human granzyme B ELISA standard, and standard reconstitution buffer. This matche 1 * 576 Tests
Catalog # ANTIA270391-696
Supplier:  ANTIBODIES.COM
New Product