Order Entry
Sweden
ContactUsLinkComponent
Human Thioredoxin/TRX Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detecti on antibody, Streptavidin-HRP, recombinant human thioredoxin-1 ELISA standard, and standard reconstitution buffer. Th 1 * 1.920 Tests
  ANTIA270504-2096
New Product
Human Thioredoxin/TRX Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detecti on antibody, Streptavidin-HRP, recombinant human thioredoxin-1 ELISA standard, and standard reconstitution buffer. Th 1 * 1.920 Tests
  ANTIA270504-2096
New Product