Order Entry
Sweden
ContactUsLinkComponent
Human IgG Matched Antibody Pair Kit (High Sensitivity) includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, native human IgG ELISA standard, and standard reconstitution buffer. This matched 1 * 576 Tests
Catalog # ANTIA270396-696
Supplier:  ANTIBODIES.COM
New Product
Human IgG Matched Antibody Pair Kit (High Sensitivity) includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, native human IgG ELISA standard, and standard reconstitution buffer. This matched 1 * 576 Tests
Catalog # ANTIA270396-696
Supplier:  ANTIBODIES.COM
New Product