Order Entry
Sweden
ContactUsLinkComponent
Mouse IL-6 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant mouse IL-6 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 576 Tests
Catalog # ANTIA270446-696
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-6 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant mouse IL-6 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 576 Tests
Catalog # ANTIA270446-696
Supplier:  ANTIBODIES.COM
New Product