Order Entry
Sweden
ContactUsLinkComponent
Bovine IL-8 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant IL-8 ELISA standard, and standard reconstitution buffer. This matched antibody pair k 1 * 576 Tests
Catalog # ANTIA270376-696
Supplier:  Antibodies.com
New Product
Bovine IL-8 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant IL-8 ELISA standard, and standard reconstitution buffer. This matched antibody pair k 1 * 576 Tests
Catalog # ANTIA270376-696
Supplier:  Antibodies.com