Rat IL-22 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, recombinant mouse IL-22 ELISA standard, and standard reconstitution buffer. This matched antibody d 1 * 576 Tests
Catalog # ANTIA270538-696
Supplier: ANTIBODIES.COM
New Product