Order Entry
Sweden
ContactUsLinkComponent
Mouse IgE Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, native mouse IgE ELISA standard, and standard reconstitution buffer. This matched antibody pair kit 1 * 576 Tests
Catalog # ANTIA270433-696
Supplier:  ANTIBODIES.COM
New Product
Mouse IgE Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, native mouse IgE ELISA standard, and standard reconstitution buffer. This matched antibody pair kit 1 * 576 Tests
Catalog # ANTIA270433-696
Supplier:  ANTIBODIES.COM
New Product