Mouse IL-4 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-HRP, recombinant mouse IL-4 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
Catalog # ANTIA270526-696
Supplier: ANTIBODIES.COM
New Product