Order Entry
Sweden
ContactUsLinkComponent
Human Granzyme A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-HRP, recombinant human granzyme A ELISA standard, and standard reconstitution buffer. This match 1 * 1.920 Tests
  ANTIA270476-2096
New Product
Human Granzyme A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-HRP, recombinant human granzyme A ELISA standard, and standard reconstitution buffer. This match 1 * 1.920 Tests
  ANTIA270476-2096
New Product