Order Entry
Sweden
ContactUsLinkComponent
Monkey IL-8 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human IL-8 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 1.920 Tests
Catalog # ANTIA270513-2096
Supplier:  ANTIBODIES.COM
New Product
Monkey IL-8 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human IL-8 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 1.920 Tests
Catalog # ANTIA270513-2096
Supplier:  ANTIBODIES.COM
New Product