Order Entry
Sweden
ContactUsLinkComponent
Human Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detect ion antibody, Streptavidin-AP, recombinant human Interferon gamma ELISA standard, and standard reconstitution buffer. 1 * 576 Tests
Catalog # ANTIA270393-696
Supplier:  ANTIBODIES.COM
New Product
Human Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detect ion antibody, Streptavidin-AP, recombinant human Interferon gamma ELISA standard, and standard reconstitution buffer. 1 * 576 Tests
Catalog # ANTIA270393-696
Supplier:  ANTIBODIES.COM
New Product