Order Entry
Sweden
ContactUsLinkComponent
Sheep IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched anti 1 * 1.920 Tests
Catalog # ANTIA270539-2096
Supplier:  ANTIBODIES.COM
New Product
Sheep IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched anti 1 * 1.920 Tests
Catalog # ANTIA270539-2096
Supplier:  ANTIBODIES.COM
New Product