Order Entry
Sweden
ContactUsLinkComponent
Horse IL-10 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human IL-10 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 576 Tests
Catalog # ANTIA325373-696
Supplier:  ANTIBODIES.COM
New Product
Horse IL-10 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human IL-10 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 576 Tests
Catalog # ANTIA325373-696
Supplier:  ANTIBODIES.COM
New Product