Order Entry
Sweden
ContactUsLinkComponent
Goat IL-5 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, recombinant bovine IL-5 ELISA standard, and standard reconstitution buffer. This matched antibody d 1 * 1.920 Tests
  ANTIA285926-2096
New Product
Goat IL-5 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, recombinant bovine IL-5 ELISA standard, and standard reconstitution buffer. This matched antibody d 1 * 1.920 Tests
  ANTIA285926-2096
New Product