Order Entry
Sweden
ContactUsLinkComponent
Monkey MCP1 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human CCL2 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
Catalog # ANTIA325336-696
Supplier:  ANTIBODIES.COM
New Product
Monkey MCP1 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human CCL2 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
Catalog # ANTIA325336-696
Supplier:  ANTIBODIES.COM
New Product