Order Entry
Sweden
ContactUsLinkComponent
Horse IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, native equine IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair ki 1 * 576 Tests
Katalog # ANTIA325378-696
Supplier:  ANTIBODIES.COM
New Product
Horse IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, native equine IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair ki 1 * 576 Tests
Katalog # ANTIA325378-696
Supplier:  ANTIBODIES.COM
New Product