Order Entry
Sweden
ContactUsLinkComponent
Human Interferon gamma ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro quantitative determination of huma n Interferon gamma in culture medium of lymphocytes stimulated with antigen or mitogen in vitro. 1 * 96 Tests
Catalog # ANTIA324362-96
Supplier:  Antibodies.com
New Product
Human Interferon gamma ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro quantitative determination of huma n Interferon gamma in culture medium of lymphocytes stimulated with antigen or mitogen in vitro. 1 * 96 Tests
Catalog # ANTIA324362-96
Supplier:  Antibodies.com