Order Entry
Sweden
ContactUsLinkComponent
Human MDC Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an biotin-labelled monoclonal detection antib ody, Streptavidin-HRP, recombinant human CCL22 ELISA standard, and standard reconstitution buffer A5. This matched an 1 * 1.920 Tests
  ANTIA325347-2096
New Product
Human MDC Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an biotin-labelled monoclonal detection antib ody, Streptavidin-HRP, recombinant human CCL22 ELISA standard, and standard reconstitution buffer A5. This matched an 1 * 1.920 Tests
  ANTIA325347-2096
New Product