Bovine TNF alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-HRP, recombinant bovine TNF alpha ELISA standard, and standard reconstitution buffer. This match 1 * 1.920 Tests
Catalog # ANTIA325324-2096
Supplier: ANTIBODIES.COM
New Product