Order Entry
Sweden
ContactUsLinkComponent
Rabbit Haptoglobin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of rabbit Haptoglobin in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samp 1 * 96 Tests
Catalog # ANTIA326618-96
Supplier:  Antibodies.com
New Product
Rabbit Haptoglobin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of rabbit Haptoglobin in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samp 1 * 96 Tests
Catalog # ANTIA326618-96
Supplier:  Antibodies.com