Order Entry
Sweden
ContactUsLinkComponent
Horse IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, native equine IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair k 1 * 576 Tests
  ANTIA325379-696
New Product
Horse IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, native equine IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair k 1 * 576 Tests
  ANTIA325379-696
New Product