Order Entry
Sweden
ContactUsLinkComponent
Human C2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human C2 in serum, plasma or other biological fluids. 1 * 48 Tests
Katalog # ANTIA3189-48
Supplier:  ANTIBODIES.COM
New Product
Human C2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human C2 in serum, plasma or other biological fluids. 1 * 48 Tests
Katalog # ANTIA3189-48
Supplier:  ANTIBODIES.COM
New Product