Order Entry
Sweden
ContactUsLinkComponent
Bovine IL-5 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant bovine IL-5 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
Catalog # ANTIA285921-696
Supplier:  Antibodies.com
New Product
Bovine IL-5 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant bovine IL-5 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
Catalog # ANTIA285921-696
Supplier:  Antibodies.com