Order Entry
Sweden
ContactUsLinkComponent
Human MCP1 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-HRP, recombinant human CCL2 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 1.920 Tests
Catalog # ANTIA325334-2096
Supplier:  ANTIBODIES.COM
New Product
Human MCP1 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-HRP, recombinant human CCL2 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 1.920 Tests
Catalog # ANTIA325334-2096
Supplier:  ANTIBODIES.COM
New Product