Order Entry
Sweden
ContactUsLinkComponent
Monkey MDC Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an biotin-labelled monoclonal detection anti body, Streptavidin-HRP, recombinant human CCL22 ELISA standard, and standard reconstitution buffer A5. This matched a 1 * 576 Tests
Catalog # ANTIA325349-696
Supplier:  ANTIBODIES.COM
New Product
Monkey MDC Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an biotin-labelled monoclonal detection anti body, Streptavidin-HRP, recombinant human CCL22 ELISA standard, and standard reconstitution buffer A5. This matched a 1 * 576 Tests
Catalog # ANTIA325349-696
Supplier:  ANTIBODIES.COM
New Product