Order Entry
Sweden
ContactUsLinkComponent
Monkey Granzyme B Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection a ntibody, Streptavidin-AP, recombinant monkey granzyme B ELISA standard, and standard reconstitution buffer. This matc 1 * 576 Tests
Catalog # ANTIA325358-696
Supplier:  ANTIBODIES.COM
New Product
Monkey Granzyme B Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection a ntibody, Streptavidin-AP, recombinant monkey granzyme B ELISA standard, and standard reconstitution buffer. This matc 1 * 576 Tests
Catalog # ANTIA325358-696
Supplier:  ANTIBODIES.COM
New Product