0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Salbutamol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Salbutamol in urin e. 1 * 40 Tests
Catalog # ANTIA327245-40
Supplier:
ANTIBODIES.COM
New Product
Expand image
Salbutamol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Salbutamol in urin e. 1 * 40 Tests
Catalog # ANTIA327245-40
Supplier:
ANTIBODIES.COM
New Product