Order Entry
Sweden
ContactUsLinkComponent
Neomycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Neomycin in raw comm ingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327228-96
Supplier:  ANTIBODIES.COM
New Product
Neomycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Neomycin in raw comm ingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327228-96
Supplier:  ANTIBODIES.COM
New Product