Order Entry
Sweden
ContactUsLinkComponent
Aflatoxin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Aflatoxin in grain, feed, oil sample. 1 * 50 Tests
Catalog # ANTIA327175-50
Supplier:  ANTIBODIES.COM
New Product
Aflatoxin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Aflatoxin in grain, feed, oil sample. 1 * 50 Tests
Catalog # ANTIA327175-50
Supplier:  ANTIBODIES.COM
New Product